Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST6GALNAC6
Entrez Gene ID
101123278
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
Gene Aliases
-
Chromosome Location
Chr.3: 8578953 - 8595768 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005616.4 XP_004005665.1 4 - 5 427 100
XM_012162328.3 XP_012017718.1 3 - 4 374 100
XM_012162341.3 XP_012017731.1 3 - 4 546 100
XM_027966408.1 XP_027822209.1 4 - 5 789 100
XM_027966409.1 XP_027822210.1 4 - 5 471 100
XM_027966410.1 XP_027822211.1 4 - 5 591 100
XM_027966411.1 XP_027822212.1 3 - 4 395 100
XM_027966412.1 XP_027822213.1 3 - 4 350 100

View Full Product Details