Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMA8
Entrez Gene ID
101117634
Gene Name
proteasome 20S subunit alpha 8
Gene Aliases
-
Chromosome Location
Chr.23: 35414601 - 35436271 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020462.4 XP_004020511.1 2 - 3 524 113

View Full Product Details