Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UXT
Entrez Gene ID
101114935
Gene Name
ubiquitously expressed prefoldin like chaperone
Gene Aliases
-
Chromosome Location
Chr.X: 59464543 - 59471405 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022128.4 XP_004022177.1 3 - 4 294 99

View Full Product Details