Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL44
Entrez Gene ID
101121751
Gene Name
mitochondrial ribosomal protein L44
Gene Aliases
-
Chromosome Location
Chr.2: 241108831 - 241117052 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012147113.3 XP_012002503.2 2 - 3 646 98

View Full Product Details