Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STON1
Entrez Gene ID
105601863
Gene Name
stonin 1
Gene Aliases
-
Chromosome Location
Chr.3: 81153598 - 81213579 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966864.1 XP_027822665.1 2 - 3 2045 108

View Full Product Details