Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHDC9
Entrez Gene ID
101101852
Gene Name
kelch domain containing 9
Gene Aliases
-
Chromosome Location
Chr.1: 119628847 - 119630722 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002681.3 XP_004002730.2 1 - 2 618 91

View Full Product Details