Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAT4
Entrez Gene ID
101123237
Gene Name
FAT atypical cadherin 4
Gene Aliases
-
Chromosome Location
Chr.17: 36034380 - 36221795 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027956286.1 XP_027812087.1 14 - 15 12650 75

View Full Product Details