Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPIC
Entrez Gene ID
101111017
Gene Name
peptidylprolyl isomerase C
Gene Aliases
-
Chromosome Location
Chr.5: 30064237 - 30078423 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008676.4 XP_004008725.2 1 - 2 195 104

View Full Product Details