Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS19
Entrez Gene ID
101106101
Gene Name
ribosomal protein S19
Gene Aliases
-
Chromosome Location
Chr.14: 54524442 - 54532381 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015294.3 XP_004015343.1 4 - 5 827 106
XM_027978401.1 XP_027834202.1 4 - 5 374 106

View Full Product Details