Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYRM4
Entrez Gene ID
101109130
Gene Name
LYR motif containing 4
Gene Aliases
-
Chromosome Location
Chr.20: 52878599 - 52966888 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019146.4 XP_004019195.1 2 - 3 312 104

View Full Product Details