Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BSCL2
Entrez Gene ID
101113634
Gene Name
BSCL2 lipid droplet biogenesis associated, seipin
Gene Aliases
-
Chromosome Location
Chr.21: 42877670 - 42893152 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012102042.2 XP_011957432.1 10 - 11 1374 105
XM_012102044.3 XP_011957434.1 10 - 11 1389 105
XM_012102045.3 XP_011957435.1 10 - 11 1408 105
XM_012102047.2 XP_011957437.1 10 - 11 1740 105
XM_012102048.3 XP_011957438.1 9 - 10 1142 105
XM_012102049.3 XP_011957439.1 9 - 10 1234 105
XM_027960097.1 XP_027815898.1 10 - 11 1372 105
XM_027960098.1 XP_027815899.1 10 - 11 1369 105
XM_027960099.1 XP_027815900.1 10 - 11 1363 105

View Full Product Details