Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAC3D1
Entrez Gene ID
101105453
Gene Name
SAC3 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.21: 45057318 - 45060521 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019661.4 XP_004019710.2 2 - 3 735 73
XM_015103175.2 XP_014958661.2 2 - 3 794 73

View Full Product Details