Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TUBB6
Entrez Gene ID
101106224
Gene Name
tubulin beta 6 class V
Gene Aliases
-
Chromosome Location
Chr.23: 48287650 - 48296399 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961046.1 XP_027816847.1 3 - 4 353 62

View Full Product Details