Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC24
Entrez Gene ID
101108713
Gene Name
tetratricopeptide repeat domain 24
Gene Aliases
-
Chromosome Location
Chr.1: 112699268 - 112710674 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015092305.2 XP_014947791.2 3 - 4 1001 84
XM_027976425.1 XP_027832226.1 3 - 4 1001 84
XM_027976429.1 XP_027832230.1 3 - 4 1001 84
XM_027976433.1 XP_027832234.1 3 - 4 1001 84

View Full Product Details