Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALKBH7
Entrez Gene ID
101109968
Gene Name
alkB homolog 7
Gene Aliases
-
Chromosome Location
Chr.5: 16603336 - 16604979 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015095865.2 XP_014951351.2 2 - 3 403 70

View Full Product Details