Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL10
Entrez Gene ID
101105498
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
-
Chromosome Location
Chr.5: 38097619 - 38106968 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015095967.2 XP_014951453.2 2 - 3 457 91

View Full Product Details