Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIAH1
Entrez Gene ID
101111116
Gene Name
siah E3 ubiquitin protein ligase 1
Gene Aliases
-
Chromosome Location
Chr.14: 18183535 - 18210882 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004014973.3 XP_004015022.3 1 - 2 585 76

View Full Product Details