Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPK2
Entrez Gene ID
101104600
Gene Name
uroplakin 2
Gene Aliases
-
Chromosome Location
Chr.15: 32359289 - 32361529 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979002.1 XP_027834803.1 4 - 5 463 62

View Full Product Details