Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIXL1
Entrez Gene ID
101105839
Gene Name
Mix paired-like homeobox
Gene Aliases
-
Chromosome Location
Chr.12: 31925484 - 31927629 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004023279.3 XP_004023328.2 1 - 2 411 71

View Full Product Details