Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A14
Entrez Gene ID
101114976
Gene Name
membrane spanning 4-domains A14
Gene Aliases
-
Chromosome Location
Chr.15: 89509548 - 89536084 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979927.1 XP_027835728.1 1 - 2 135 76

View Full Product Details