Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBX4
Entrez Gene ID
101110952
Gene Name
chromobox 4
Gene Aliases
-
Chromosome Location
Chr.11: 10555177 - 10561265 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974232.1 XP_027830033.1 4 - 5 279 76

View Full Product Details