Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPRED3
Entrez Gene ID
101112083
Gene Name
sprouty related EVH1 domain containing 3
Gene Aliases
-
Chromosome Location
Chr.14: 51082453 - 51092270 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978238.1 XP_027834039.1 2 - 3 1762 90

View Full Product Details