Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIGH
Entrez Gene ID
101114362
Gene Name
phosphatidylinositol glycan anchor biosynthesis class H
Gene Aliases
-
Chromosome Location
Chr.7: 82976889 - 82983413 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010735.4 XP_004010784.1 2 - 3 449 99

View Full Product Details