Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA12
Entrez Gene ID
101118733
Gene Name
serpin family A member 12
Gene Aliases
-
Chromosome Location
Chr.18: 59552176 - 59567795 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012099001.3 XP_011954391.1 4 - 5 1216 76
XM_012099002.3 XP_011954392.1 4 - 5 1393 76

View Full Product Details