Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LEKR1
Entrez Gene ID
101120544
Gene Name
leucine, glutamate and lysine rich 1
Gene Aliases
-
Chromosome Location
Chr.1: 251266796 - 251496119 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012100804.3 XP_011956194.1 11 - 12 1479 186

View Full Product Details