Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SBSN
Entrez Gene ID
101120596
Gene Name
suprabasin
Gene Aliases
-
Chromosome Location
Chr.14: 48896626 - 48901568 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977407.1 XP_027833208.1 1 - 2 1845 60

View Full Product Details