Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARID3C
Entrez Gene ID
101116967
Gene Name
AT-rich interaction domain 3C
Gene Aliases
-
Chromosome Location
Chr.2: 38910298 - 38917674 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963899.1 XP_027819700.1 3 - 4 576 63

View Full Product Details