Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPEM1
Entrez Gene ID
101120848
Gene Name
spermatid maturation 1
Gene Aliases
-
Chromosome Location
Chr.11: 36251294 - 36252994 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974978.1 XP_027830779.1 1 - 2 727 78

View Full Product Details