Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC114110825
Entrez Gene ID
114110825
Gene Name
tyrosine-protein phosphatase non-receptor type 20-like
Gene Aliases
-
Chromosome Location
Chr.25: 44378882 - 44395106 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962280.1 XP_027818081.1 3 - 4 607 102

View Full Product Details