Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLAG1
Entrez Gene ID
101115630
Gene Name
PLAG1 zinc finger
Gene Aliases
-
Chromosome Location
Chr.9: 39062460 - 39075210 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012183720.2 XP_012039110.1 2 - 3 1501 90

View Full Product Details