Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNB3
Entrez Gene ID
101122833
Gene Name
cholinergic receptor nicotinic beta 3 subunit
Gene Aliases
-
Chromosome Location
Chr.26: 40825114 - 40855576 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012106019.3 XP_011961409.1 1 - 2 53 74

View Full Product Details