Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC101115225
Entrez Gene ID
101115225
Gene Name
sialic acid-binding Ig-like lectin 8
Gene Aliases
-
Chromosome Location
Chr.14: 59719177 - 59721339 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977946.1 XP_027833747.1 2 - 3 533 91

View Full Product Details