Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLLT3
Entrez Gene ID
101102874
Gene Name
MLLT3 super elongation complex subunit
Gene Aliases
-
Chromosome Location
Chr.2: 95894301 - 96172405 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004397.3 XP_004004446.2 10 - 11 1835 97

View Full Product Details