Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS18
Entrez Gene ID
100036761
Gene Name
ribosomal protein S18
Gene Aliases
-
Chromosome Location
Chr.20: 9148687 - 9153444 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018745.3 XP_004018794.1 5 - 6 462 79

View Full Product Details