Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB12
Entrez Gene ID
101103204
Gene Name
RAB12, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.23: 46270831 - 46292166 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015103654.2 XP_014959140.2 4 - 5 555 102

View Full Product Details