Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AREG
Entrez Gene ID
443427
Gene Name
amphiregulin
Gene Aliases
-
Chromosome Location
Chr.6: 99250625 - 99260260 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012180163.3 XP_012035553.2 5 - 6 948 100

View Full Product Details