Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT39
Entrez Gene ID
100526786
Gene Name
keratin 39
Gene Aliases
-
Chromosome Location
Chr.11: 22016383 - 22023827 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012186079.3 XP_012041469.2 4 - 5 752 109

View Full Product Details