Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRSS33
Entrez Gene ID
105604615
Gene Name
serine protease 33
Gene Aliases
-
Chromosome Location
Chr.24: 2212621 - 2216190 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962106.1 XP_027817907.1 4 - 5 527 70

View Full Product Details