Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A4
Entrez Gene ID
101110981
Gene Name
solute carrier family 16 member 4
Gene Aliases
-
Chromosome Location
Chr.1: 92234773 - 92266580 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002305.4 XP_004002354.3 3 - 4 398 105

View Full Product Details