Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAZALD1
Entrez Gene ID
101112689
Gene Name
Kazal type serine peptidase inhibitor domain 1
Gene Aliases
-
Chromosome Location
Chr.22: 24509632 - 24514705 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960353.1 XP_027816154.1 2 - 3 712 82

View Full Product Details