Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFA7
Entrez Gene ID
101103062
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
Gene Aliases
-
Chromosome Location
Chr.5: 15278124 - 15285421 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008565.3 XP_004008614.1 1 - 2 92 77

View Full Product Details