Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNG4
Entrez Gene ID
101102663
Gene Name
potassium voltage-gated channel modifier subfamily G member 4
Gene Aliases
-
Chromosome Location
Chr.14: 11176008 - 11191481 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977584.1 XP_027833385.1 2 - 3 871 92

View Full Product Details