Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAPLN3
Entrez Gene ID
101102761
Gene Name
hyaluronan and proteoglycan link protein 3
Gene Aliases
-
Chromosome Location
Chr.18: 17878541 - 17897885 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017742.3 XP_004017791.1 2 - 3 1690 63

View Full Product Details