Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO2
Entrez Gene ID
101112079
Gene Name
F-box protein 2
Gene Aliases
-
Chromosome Location
Chr.12: 45433746 - 45439672 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975641.1 XP_027831442.1 1 - 2 3923 67

View Full Product Details