Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR4
Entrez Gene ID
101104156
Gene Name
HEAT repeat containing 4
Gene Aliases
-
Chromosome Location
Chr.7: 88681981 - 88721531 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971998.1 XP_027827799.1 11 - 12 4614 67

View Full Product Details