Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A3GALT2
Entrez Gene ID
101103704
Gene Name
alpha 1,3-galactosyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.2: 249986437 - 249997398 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093608.2 XP_014949094.2 2 - 3 1762 64

View Full Product Details