Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KY
Entrez Gene ID
101113821
Gene Name
kyphoscoliosis peptidase
Gene Aliases
-
Chromosome Location
Chr.1: 277388455 - 277438470 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960210.1 XP_027816011.1 9 - 10 918 85

View Full Product Details