Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAML2
Entrez Gene ID
101102586
Gene Name
mastermind like transcriptional coactivator 2
Gene Aliases
-
Chromosome Location
Chr.15: 15261143 - 15669217 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015100757.2 XP_014956243.2 2 - 3 3427 92

View Full Product Details