Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC114116593
- Entrez Gene ID
-
114116593
- Gene Name
- U3 small nucleolar RNA-associated protein 14 homolog A
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 9871033 - 9873564 on Build Oar_rambouillet_v1.0
- Gene Symbol
- UTP14A
- Entrez Gene ID
-
101120809
- Gene Name
- UTP14A small subunit processome component
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 121582253 - 121599131 on Build Oar_rambouillet_v1.0
View Full Product Details