Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP6V0E1
Entrez Gene ID
101108422
Gene Name
ATPase H+ transporting V0 subunit e1
Gene Aliases
-
Chromosome Location
Chr.16: 5014612 - 5042749 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016871.3 XP_004016920.1 2 - 3 261 105

View Full Product Details